

2-Chloro-6-(trifluoromethyl)aniline ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:433-94-3
:
Formula:C₇H₅ClF₃N
MDL Number:MFCD00272565
:195.57 g/mol
Boiling Pt:39…40 °C (0.1 mmHg)
Density:1.42
Storage Temperature:Ambient
EINECS:207-091-8
UN:2810
ADR:6.1,III
:GHS06
:GHS07
Frequently Bought Together