
4-Bromo-m-cresol 98%
Catalog # ACRO368200050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:57 to 61 °C
- GC:≥97.5 %
Specification Test Results
About this item
:
:187.04 g/mol
Boiling Pt:142…145 °C (23 mmHg)
Melting Pt:57…61 °C
EINECS:238-464-3
:GHS07
CAS Number:14472-14-1
Formula:C₇H₇BrO
MDL Number:MFCD00079723