

3-Ethynylaniline 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:54060-30-9
:
Formula:C₈H₇N
MDL Number:MFCD00014779
:117.15 g/mol
Boiling Pt:92…93 °C (2 mmHg)
Melting Pt:27 °C
Density:1.04 g/cm³
Flash Pt:59 °C
Storage Temperature:Refrigerator
EINECS:258-944-6
UN:1993
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6140 to 1.6180 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together