
cis-Stilbene ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:645-49-8
:
Formula:(C₆H₅)CHCH(C₆H₅)
MDL Number:MFCD00004788
:180.25 g/mol
Merck Index:14,8817
Boiling Pt:307 °C
Melting Pt:1…2 °C
Density:1.04
Flash Pt:>110 °C (230 °F)
EINECS:211-445-7
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together