Order Entry
Italy
ContactUsLinkComponent
1,2,4-Trihydroxybenzene for synthesis, Sigma-Aldrich®
1,2,4-Trihydroxybenzene for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
1,2,4-Trihydroxybenzene for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    137 - 140 °C
  • Molar Mass:
    126.11 g/mol
  • Density:
    1.45 g/cm3 (20 °C)
  • Hill Formula:
    C6H6O3

Specification Test Results

Related Information

Frequently Bought Together