

Pyrogallol 99% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:87-66-1
:
Formula:C₆H₆O₃
MDL Number:MFCD00002192
:126.11 g/mol
Merck Index:14,8000
Boiling Pt:309 °C
Melting Pt:131…135 °C
Density:1.45
Storage Temperature:Ambient
EINECS:201-762-9
UN:2811
ADR:6.1,III
:GHS08
:GHS07
Related Information
Frequently Bought Together