

Resorcinol for synthesis, Sigma-Aldrich®
: Merck
:108-46-3
:
:C₆H₆O₂
:MFCD00002269
:12,08323
:281 °C (1013 hPa)
:109…111 °C
:1.272 g/cm³ (20 °C)
:127 °C (closed cup)
:203-585-2
:2876
:6.1,III
:M-Clarity™ quality level = MQ200
:110.11 g/mol
:GHS07
:GHS09
:Ambient
- Solubility:1000 g/l (20 °C)
- Melting Point:109 - 111 °C
- Molar Mass:110.11 g/mol
- Bulk Density:600 - 700 kg/m3
- Boiling Point:281 °C (1013 hPa)
- Vapor Pressure:0.1 hPa (20 °C)
- Flash Point:171 °C
- Explosion Limit:1.4 %(V)
- Density:1.28 g/cm3 (20 °C)
- pH:4 - 6 (100 g/l, H2O, 20 °C)
- Ignition Point:605 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Frequently Bought Together