Order Entry
Italy
ContactUsLinkComponent
o-Anisidine 99+%
o-Anisidine 99+%
Certificates
undefined
o-Anisidine 99+%

Specification Test Results

  • Appearance:
    Clear colorless to yellow-brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5730 to 1.5755 (20°C, 589 nm)
  • GC:
    ≥98.0 %

Specification Test Results

Related Information

Frequently Bought Together