Order Entry
Italy
ContactUsLinkComponent
 
 
2-Fluorobenzenesulphonyl chloride 97%
 
undefined
 
2-Fluorobenzenesulphonyl chloride 97%

 

  • Appearance:
    Clear colorless to yellow-brown Liquid or solid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5350 to 1.5390 (20°C, 589 nm)
  • Titration Argentometric:
    ≥96.0 % (ex Cl)

 

 

Frequently Bought Together