
2,4-Difluorophenyl isothiocyanate 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:141106-52-7
:
Formula:C₇H₃F₂NS
MDL Number:MFCD00041044
:171.17 g/mol
Boiling Pt:98…99 °C (30 mmHg)
Density:1.349
Flash Pt:92 °C (197 °F)
Storage Temperature:Ambient
EINECS:624-035-1
ADR:,
:GHS07
Related Information
Frequently Bought Together