
2,6-Difluorobenzoic acid
Catalog # EHERC12632010
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:158.1 g/mol
Boiling Pt:72…77 °C (13 mmHg)
Melting Pt:159…160 °C
EINECS:206-856-3
:GHS07
Storage Temperature:Ambient
CAS Number:385-00-2
Formula:C₇H₄F₂O₂
MDL Number:MFCD00002411