Order Entry
Italy
ContactUsLinkComponent
Propyl 4-hydroxybenzoate sodium salt
Corrosive
Propyl 4-hydroxybenzoate sodium salt
Catalog # PROOMM0432.01
CAS Number:  
Corrosive
Propyl 4-hydroxybenzoate sodium salt
Catalog # PROOMM0432.01
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :202.19 g/mol
EINECS:252-488-1
 :GHS05
Storage Temperature:Ambient
CAS Number:35285-69-9
Formula:C₁₀H₁₁NaO₃
MDL Number:MFCD00016477