Ethyl-4-nitrobenzoate
Glass bottle for solids, 100 mg
Catalog # PROOMM0094.04
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: Ethyl 4-nitrobenzoate, 4-Nitrobenzoic acid ethyl ester
- CAS Number:99-77-4
- Formula:C₉H₉NO₄
- MDL Number:MFCD00007351
- Molecular Weight:195.17 g/mol
- Storage Temperature:Ambient
- EINECS:202-786-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



