3,4-Dihydroxybenzhydrazide ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 39635-11-5 Molecular Formula: C7H8N2O3 Formula Weight: 168.15 Melting Point: ca 265° dec. Beilstein Reference: 2642968
Synonyms: 3,4-Dihydroxybenzoic hydrazide, 3,4-Dihydroxybenzoic acid hydrazide, 3,4-Dihydroxybenzene-1-carbohydrazide, 3,4-Dihydroxybenzohydrazide
- CAS Number:39635-11-5
- Formula:C₇H₈N₂O₃
- MDL Number:MFCD00017064
- Molecular Weight:168.15 g/mol
- Melting Pt:∼ 265 °C (dec.)
- Storage Temperature:Ambiente
- EINECS:254-550-3
2 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 39635-11-5 Molecular Formula: C7H8N2O3 Formula Weight: 168.15 Melting Point: ca 265° dec. Beilstein Reference: 2642968
Synonyms: 3,4-Dihydroxybenzoic hydrazide, 3,4-Dihydroxybenzoic acid hydrazide, 3,4-Dihydroxybenzene-1-carbohydrazide, 3,4-Dihydroxybenzohydrazide
- CAS Number:39635-11-5
- Formula:C₇H₈N₂O₃
- MDL Number:MFCD00017064
- Molecular Weight:168.15 g/mol
- Melting Pt:∼ 265 °C (dec.)
- Storage Temperature:Ambiente
- EINECS:254-550-3
Documents
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



