Order Entry
Italy
ContactUsLinkComponent
4-Nitrobenzoyl chloride for synthesis, Sigma-Aldrich®
Corrosive
4-Nitrobenzoyl chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
4-Nitrobenzoyl chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    73 °C
  • Molar Mass:
    185.56 g/mol
  • Boiling Point:
    155 °C (20 hPa)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together