Order Entry
Italy
ContactUsLinkComponent
3-Nitrobenzoyl chloride 98%
Irritant
Corrosive
3-Nitrobenzoyl chloride 98%
Catalog # ACRO155840050
CAS Number:  
undefined
Irritant
Corrosive
3-Nitrobenzoyl chloride 98%
Catalog # ACRO155840050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Brown Low melting solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    30 to 35 °C
  • Titration RCOCl:
    ≥97.5 %
  • UV:
    at 300 nm A: ≤0.3
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0005 %
  • Acidity:
    ≤0.0003 meq/g
  • Alkalinity:
    ≤0.0005 meq/g

Specification Test Results

About this item

 :
 :185.57 g/mol
Boiling Pt:275 °C (1013 hPa)
Melting Pt:30…35 °C
Density:1.42 g/cm³
Flash Pt:>112 °C
Storage Temperature:Ambient
EINECS:204-505-9
UN:1759
ADR:8,II
 :GHS07
 :GHS05
CAS Number:121-90-4
Formula:C₇H₄ClNO₃
MDL Number:MFCD00007247