Isophthalonitrile for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:626-17-5
:
Formula:C₈H₄N₂
MDL Number:MFCD00001795
:128.13 g/mol
Boiling Pt:288 °C (1013 hPa)
Melting Pt:158…160 °C
Density:4.42 g/cm³ (20 °C)
Flash Pt:>150 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,II
:M-Clarity™ quality level = MQ200
EINECS:210-933-7
Specification Test Results
- Solubility:0.7 g/l (20 °C)
- Melting Point:162 - 163 °C
- Molar Mass:128.13 g/mol
- Bulk Density:500 kg/m3
- Boiling Point:288 °C (1013 hPa)
- Vapor Pressure:0.014 hPa (20 °C)
- Density:1.3 g/cm3 (20 °C)
- Synonyms:Benzene-1,3-dicarbonitrile, 1,3-Dicyanobenzene
- Hill Formula:C8H4N2
Specification Test Results
Frequently Bought Together