3-Chlorophenyl isocyanate
Catalog # 8.02602.0250
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:250 ml
- Seq:2590
Specifications
About this item
:
:153.57 g/mol
Boiling Pt:202 °C (1013 hPa)
Melting Pt:–4.4 °C
Density:1.26 g/cm³ (20 °C)
EINECS:220-822-5
UN:3382
ADR:6.1,I
CAS Number:2909-38-8
Formula:C₇H₄ClNO
MDL Number:MFCD00002016