Order Entry
Italy
ContactUsLinkComponent
2-Chlorobenzoyl chloride for synthesis, Sigma-Aldrich®
Corrosive
Irritant
2-Chlorobenzoyl chloride for synthesis, Sigma-Aldrich®
Catalog # 8.21989.0250
Supplier:  Merck
CAS Number:  
Corrosive
Irritant
2-Chlorobenzoyl chloride for synthesis, Sigma-Aldrich®
Catalog # 8.21989.0250
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 ml
  • Seq:
    2590

Specifications

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    -4 °C
  • Molar Mass:
    175.01 g/mol
  • Boiling Point:
    225 °C (1013 hPa)
  • Vapor Pressure:
    0.1 hPa (20 °C)
  • Flash Point:
    124 °C
  • Refractive Index:
    1.5718 (20 °C, 589 nm)
  • Explosion Limit:
    1.5 - 9.4 %(V)
  • Density:
    1.38 g/cm3 (20 °C)
  • pH:
    (H2O) acidic
  • Ignition Point:
    600 °C
  • Viscosity kinematic:
    1.89 mm2/s (20 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

About this item

 :
 :175.01 g/mol
Boiling Pt:238 °C (1013 hPa)
Melting Pt:–4 °C
Density:1.38 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-194-0
UN:3265
ADR:8,II
 :M-Clarity™ quality level = MQ200
 :GHS05
 :GHS07
CAS Number:609-65-4
Formula:C₇H₄Cl₂O
MDL Number:MFCD00000660