
4-Bromobenzenesulphonyl chloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-58-8
:
Formula:C₆H₄BrClO₂S
MDL Number:MFCD00007437
:255.52 g/mol
Merck Index:13,01391
Boiling Pt:153 °C (15 mmHg)
Melting Pt:74…77 °C
Density:1,37 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-683-2
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to beige Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:74 to 77 °C
- Titration Argentometric:≥97.5 % (Ex CL)
Specification Test Results
Frequently Bought Together