

2-Bromophenyl isothiocyanate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13037-60-0
:
Formula:C₇H₄BrNS
MDL Number:MFCD00004799
:214.09 g/mol
Boiling Pt:256…257 °C
Melting Pt:20 °C
Density:1.591
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:2922
ADR:8,III
:GHS06
:GHS05
Related Information
Frequently Bought Together