Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-4,5-difluorophenol
2-Bromo-4,5-difluorophenol ≥97%
2-Bromo-4,5-difluorophenol ≥97%
L16716.06
:
Thermo Fisher Scientific
:
2-Bromo-4,5-difluorophenol ≥97%
L16716.06
:
Thermo Fisher Scientific
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
166281-37-4
:
:
C₆H₃BrF₂O
:
MFCD00070751
:
208.99 g/mol
:
1.829
:
GHS07
:
Ambient