

2,4-Dibromo-6-fluoroaniline ≥97%
Catalog # A14435.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Boiling Pt:277 °C (1013 hPa)
Melting Pt:62…64 °C
ADR:6.1,III
:268.91 g/mol
UN:2811
:GHS06
:GHS07
Storage Temperature:Ambient
CAS Number:141474-37-5
Formula:C₆H₄Br₂FN
MDL Number:MFCD00042230