Order Entry
Italy
ContactUsLinkComponent
4-Bromo-3-fluorobenzoic acid ≥98%
Irritant
4-Bromo-3-fluorobenzoic acid ≥98%
Catalog # B25475.06
CAS Number:  
Irritant
4-Bromo-3-fluorobenzoic acid ≥98%
Catalog # B25475.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:153556-42-4
 :
Formula:C₇H₄BrFO₂
MDL Number:MFCD00672932
 :219.01 g/mol
Melting Pt:210…213 °C
 :GHS07
Storage Temperature:Ambient