N-Nitroso-N-ethylaniline
Supplier: Cayman Chemical
Jump to:
About this item
Attenzione: Warning - this product is not for human or veterinary use.
Synonyms: N-ethyl-N-phenylnitrous amide, Ethylnitrosoaniline, Nitrosoethylaniline, NMA
- CAS Number:612-64-6
- Formula:C₈H₁₀N₂O
- MDL Number:MFCD01670254
- Molecular Weight:150.2 g/mol
- Storage Temperature:Freezer
- UN:2810
- ADR:6.1,I
4 Options Available Below
Product Details & Documents
Attenzione: Warning - this product is not for human or veterinary use.
Synonyms: N-ethyl-N-phenylnitrous amide, Ethylnitrosoaniline, Nitrosoethylaniline, NMA
- CAS Number:612-64-6
- Formula:C₈H₁₀N₂O
- MDL Number:MFCD01670254
- Molecular Weight:150.2 g/mol
- Storage Temperature:Freezer
- UN:2810
- ADR:6.1,I
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



