

1-Bromo-2,6-difluorobenzene ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64248-56-2
:
Formula:C₆H₃BrF₂
MDL Number:MFCD00009894
:192.99 g/mol
Boiling Pt:61 °C (35 mmHg)
Density:1.710
Flash Pt:53 °C (127 °F)
Storage Temperature:Ambient
EINECS:264-750-2
UN:1993
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together