Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-5-methoxybenzoyl chloride
2-Bromo-5-methoxybenzoyl chloride ≥97%
2-Bromo-5-methoxybenzoyl chloride ≥97%
2-Bromo-5-methoxybenzoyl chloride ≥97%
Catalog # A19164.88
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
Expand image
2-Bromo-5-methoxybenzoyl chloride ≥97%
Catalog # A19164.88
Supplier:
Thermo Fisher Scientific
CAS Number:
SDS
About this item
Product Details
Documentation
Product Details
Documentation