Order Entry
Italy
ContactUsLinkComponent
1,4-Naphthoquinone for synthesis, Sigma-Aldrich®
Toxic
Corrosive
Environment
1,4-Naphthoquinone for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Corrosive
Environment
1,4-Naphthoquinone for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    0.09 g/l (25 °C)
  • Melting Point:
    120 - 125 °C (sublimed)
  • Molar Mass:
    158.16 g/mol
  • Vapor Pressure:
    0.026 hPa (50 °C)
  • Flash Point:
    141 °C
  • Density:
    1.42 g/cm3 (20 °C)
  • pH:
    6.1 (10 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Frequently Bought Together