Benzimidazole for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:51-17-2
:
Formula:C₇H₆N₂
MDL Number:MFCD00005585
:118.14 g/mol
Merck Index:13,01082
Boiling Pt:360 °C (1013 hPa)
Melting Pt:170…173 °C
Density:1.23 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:200-081-4
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:170 - 173 °C
- Molar Mass:118.13 g/mol
- Boiling Point:360 °C
- Hill Formula:C7H6N2
Specification Test Results
Frequently Bought Together