Order Entry
Italy
ContactUsLinkComponent
4-Hydroxycinnamic acid ≥98% for synthesis, Sigma-Aldrich®
Irritant
4-Hydroxycinnamic acid ≥98% for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
4-Hydroxycinnamic acid ≥98% for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    215 - 220 °C
  • Molar Mass:
    164.16 g/mol

Specification Test Results

Related Information

Frequently Bought Together