Maleic anhydride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 2,5-Furanedione
CAS Number:108-31-6
:
Formula:C₄H₂O₃
MDL Number:MFCD00005518
:98.06 g/mol
Merck Index:13,05727
Boiling Pt:200 °C (1013 hPa)
Melting Pt:51…53 °C
Density:1.48 g/cm³ (20 °C)
EINECS:203-571-6
UN:2215
ADR:8,III
:M-Clarity™ quality level = MQ200
Flash Pt:103 °C
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble,(decomposition)
- Melting Point:51 - 53 °C
- Molar Mass:98.06 g/mol
- Bulk Density:700 - 800 kg/m3
- Boiling Point:202 °C (1013 hPa)
- Vapor Pressure:2.8 hPa (52.9 °C)
- Flash Point:103 °C
- Explosion Limit:1.4 - 7.1 %(V)
- Density:1.32 g/cm3 (55 °C)
- pH:0.8 (550 g/l, H2O, 20 °C) Hydrolysis
- Ignition Point:475 °C DIN 51794
- Hazard Class:8
- Hazard Class Description:Corrosive material
Specification Test Results
Frequently Bought Together