Maleic acid, EMPROVE® ESSENTIAL Ph. Eur., NF, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:110-16-7
:
Formula:C₄H₄O₄
MDL Number:MFCD00063177
:116.07 g/mol
Merck Index:13,05726
Boiling Pt:256 °C (1013 hPa)
Melting Pt:130…131 °C
Density:1.59 g/cm³ (20 °C)
Flash Pt:127 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-742-5
UN:3261
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together