Order Entry
Italy
Orders LinkContactUsLinkComponent
Maleic acid

Maleic acid

Catalog #
Irritant
Maleic acid
Catalog #
Supplier:  
CAS Number:  110-16-7
Maleic acid
Maleic acid
CAS Number:  110-16-7
Product Details & Documents
  • CAS Number:110-16-7
  • Formula:C₄H₄O₄
  • MDL Number:MFCD00063177
  • Molecular Weight:116.07 g/mol
  • Merck Index:13,05726
  • Boiling Pt:256 °C (1013 hPa)
  • Melting Pt:130…131 °C
  • Density:1.59 g/cm³ (20 °C)
  • Flash Pt:127 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:203-742-5
  • UN:3261
  • ADR:8,III