Order Entry
Italy
ContactUsLinkComponent
5,6-Epoxy-1-hexene 98%
Health
Flammable
5,6-Epoxy-1-hexene 98%
Certificates
undefined
Health
Flammable
5,6-Epoxy-1-hexene 98%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4240 to 1.4260 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together