4-(Methylthio)phenylacetic acid ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16188-55-9
:
Formula:C₉H₁₀O₂S
MDL Number:MFCD00192325
:182.24 g/mol
Melting Pt:98…102 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together