Order Entry
Italy
ContactUsLinkComponent
3-Methyl-3-pentanol
3-Methyl-3-pentanol
Catalog # EHERC15122400
Supplier:  EHRENSTORFER
CAS Number:  
3-Methyl-3-pentanol
Catalog # EHERC15122400
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :102.18 g/mol
Boiling Pt:123 °C (1013 hPa)
Melting Pt:–53 °C
Density:0.8452 g/cm³ (0 °C)
Storage Temperature:Ambient
EINECS:201-053-4
UN:1987
ADR:3,III
CAS Number:77-74-7
Formula:C₆H₁₄O
MDL Number:MFCD00004483