Glass beads
Supplier: CENTRAVER
Certificates
About this item
Chemically resistant glass balls suitable for use as mixing beads, boiling stones, or packing for distillation columns.
Frequently Bought Together
Chemically resistant glass balls suitable for use as mixing beads, boiling stones, or packing for distillation columns.