

tert-Butanol ≥99.5%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Catalog # 308250-2.5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 99.50 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.05 %
- Absorbance at 350 nm:Max. 0.01
- Absorbance at 300 nm:Max. 0.01
- Absorbance at 250 nm:Max. 0.20
- Absorbance at 230 nm:Max. 0.50
- Absorbance at 215 nm:Max. 1.00
- Explosion Limit:8 %
- Refractive Index:n20/D 1.387 (lit.)
- Vapor Pressure:44 mmHg ( 26 °C)
- Autoignition Temperature:896 °F
Specification Test Results
About this item
:
:74.12 g/mol
Merck Index:12,01577
Boiling Pt:82,2 °C (1013 hPa)
Melting Pt:25 °C
Density:0,7887 g/cm³ (20 °C)
Flash Pt:11 °C
Storage Temperature:Ambient
EINECS:200-889-7
UN:1120
ADR:3,II
:GHS07
:GHS02
CAS Number:75-65-0
Formula:(CH₃)₃COH
MDL Number:MFCD00004464