tert-Butanol, anhydrous ≥99.5%
:75-65-0
:
:(CH₃)₃COH
:MFCD00004464
:74.12 g/mol
:14,1542
:83 °C
:25 °C
:0.786
:200-889-7
:1120
:3,II
:Packaged under argon in resealable ChemSeal™ bottles.
:11 °C (52 °F)
:GHS02
:GHS07
:Ambient
Frequently Bought Together