tert-Butanol 99+% ACS
:75-65-0
:
:(CH₃)₃COH
:MFCD00004464
:74.12 g/mol
:14,1542
:83 °C
:25 °C
:0.786
:11 °C (52 °F)
:Ambient
:200-889-7
:1120
:3,II
:GHS02
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Frequently Bought Together