


Isobutanol 99+% for spectrophotometry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:78-83-1
:
Formula:(CH₃)₂CHCH₂OH
MDL Number:MFCD00004740
:74.12 g/mol
Merck Index:14,5131
Boiling Pt:107…108 °C
Melting Pt:–108 °C
Density:0.802
Flash Pt:28 °C (82 °F)
Storage Temperature:Ambient
EINECS:201-148-0
UN:1212
ADR:3,III
:GHS05
:GHS02
:GHS07
Related Information
Frequently Bought Together