Calbiochem® Human native transferrin (holo) (from plasma)
616424-100
:
- Pack type:Plastic ampoule
- Pk:100 mg
- Protein/peptide type:Native
- Source:Plasma
- Species:Human
- Form:Lyophilised
- Protein/peptide name:Transferrin (holo)
- Purity:>95%
- Endotoxin level:Low
- Formulation:Lyophilised from 150 mM NaCl, 20 mM sodium phosphate buffer, pH 7,4
Transferrin is a serum protein involved in iron binding and transport. Native holotransferrin from human plasma.