Methanesulphonic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-75-2
:
Formula:CH₃SO₃H
MDL Number:MFCD00007518
:96.11 g/mol
Merck Index:13,05981
Boiling Pt:217 °C (1013 hPa)
Melting Pt:20 °C
Density:1.48 g/cm³ (20 °C)
EINECS:200-898-6
UN:2922
ADR:8 (6.1),II
:M-Clarity™ quality level = MQ200
Flash Pt:189 °C (closed cup)
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:1000 g/l (20 °C)
- Melting Point:20 °C
- Molar Mass:96.11 g/mol
- Boiling Point:167 °C (13 hPa)
- Vapor Pressure:- 1 hPa (20 °C)
- Flash Point:189 °C
- Refractive Index:1.4303 (20 °C, 589 nm)
- Density:1.48 g/cm3 (20 °C)
- Ignition Point:500 °C
- Hazard Class:8
- Hazard Class Description:Corrosive material
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_806022
SDS
ProductPage_806022 ProductPage_806022
Product specification
Frequently Bought Together