

Methanesulphonic acid 99%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-75-2
:
Formula:CH₃SO₃H
MDL Number:MFCD00007518
:96.11 g/mol
Merck Index:13,05981
Boiling Pt:167 °C (10 mmHg)
Melting Pt:19 °C
Density:1.481 g/cm³
Flash Pt:189 °C
Storage Temperature:Ambient
EINECS:200-898-6
UN:3265
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4285 to 1.4315 (20°C, 589 nm)
- Color scale:≤150 APHA
- Titration with NaOH:≥98.5 %
Specification Test Results
Frequently Bought Together