


Methyl methanesulphonate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:66-27-3
:
Formula:C₂H₆O₃S
MDL Number:MFCD00007557
:110.13 g/mol
Merck Index:12,06170
Boiling Pt:202…203 °C (760 mmHg)
Melting Pt:20 °C
Density:1.3 g/cm³
Flash Pt:104 °C
Storage Temperature:Refrigerator
EINECS:200-625-0
UN:2810
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4130 to 1.4150 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together