Order Entry
Italy
ContactUsLinkComponent
1-Dodecanesulphonyl chloride ≥98%
Corrosive
1-Dodecanesulphonyl chloride ≥98%
Catalog # L01224.06
CAS Number:  
Corrosive
1-Dodecanesulphonyl chloride ≥98%
Catalog # L01224.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:10147-40-7
 :
Formula:C₁₂H₂₅ClO₂S
 :268.85 g/mol
Boiling Pt:126…130 °C (4 mmHg)
Melting Pt:39…42 °C
Density:1.04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-416-8
UN:3261
ADR:8,II
 :GHS05
MDL Number:MFCD00014728