



2-Mercaptoethanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:60-24-2
:
Formula:HSCH₂CH₂OH
MDL Number:MFCD00004890
Merck Index:13,05894
Boiling Pt:157 °C (1013 hPa)
Melting Pt:–100 °C
Density:1.12 g/cm³ (20 °C)
Flash Pt:68 °C
Storage Temperature:Ambient
EINECS:200-464-6
UN:2966
ADR:6.1,II
:M-Clarity™ quality level = MQ200
:78.14 g/mol
:GHS06
:GHS05
:GHS08
:GHS09
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:78.13 g/mol
- Boiling Point:154 - 161 °C (1013 hPa)
- Vapor Pressure:0.76 hPa (20 °C)
- Flash Point:68 °C
- Refractive Index:1.5006 (20 °C, 589 nm)
- Explosion Limit:2.3 - 18 %(V)
- Density:1.12 g/cm3 (20 °C)
- pH:4.5 - 6 (500 g/l, H2O, 20 °C)
- Ignition Point:295 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.05740.9025_TDS_EN ProductPage_805740
Product specification
msds_805740
SDS
Frequently Bought Together