Order Entry
Italy
ContactUsLinkComponent
 
 
Aluminium isopropoxide for synthesis, Sigma-Aldrich®
 :  Merck
 
 
Aluminium isopropoxide for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    130 - 134 °C
  • Molar Mass:
    204.24 g/mol
  • Boiling Point:
    94 °C (0.6 hPa)
  • Vapor Pressure:
    0.13 hPa (21 °C)
  • Flash Point:
    26 °C
  • Refractive Index:
    1.432 (23 °C, 589 nm)
  • Explosion Limit:
    2 %(V)
  • Density:
    1.035 g/cm3 (20 °C)
  • Hazard Class:
    4.1
  • Hazard Class Description:
    Flammable Solid

 

Frequently Bought Together