Order Entry
Italy
ContactUsLinkComponent
 
 
 
 
Potassium-tert-butoxide for synthesis, Sigma-Aldrich®
 :  Merck
 
 
 
 
Potassium-tert-butoxide for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) Hydrolysis,(rigorous decomposition)
  • Melting Point:
    256 - 258 °C
  • Molar Mass:
    112.21 g/mol
  • Bulk Density:
    500 kg/m3
  • Boiling Point:
    220 °C (1 hPa) (sublimed)
  • Vapor Pressure:
    0.07 hPa (180 °C)
  • Density:
    0.978 g/cm3 (20 °C)
  • pH:
    13 (5 g/l, H2O, 20 °C) Hydrolysis
  • Ignition Point:
    360 °C

 

Frequently Bought Together