Potassium-tert-butoxide ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:865-47-4
:
Formula:C₄H₉KO
MDL Number:MFCD00012162
:112.21 g/mol
Melting Pt:∼ 255 °C (dec.)
Storage Temperature:Ambient
EINECS:212-740-3
UN:3206
ADR:4.2,II
:GHS02
:GHS05
:GHS07
Related Information
Frequently Bought Together